Orders LinkContactUsLinkComponent
VWR® Maximum, 耐切創グローブ、
VWR® Maximum, 耐切創グローブ、
Supplier:   VWR Collection
VWR® Maximum, 耐切創グローブ、
Supplier:  VWR Collection
Certificates
VWR® Maximum, 耐切創グローブ、
6 Options Available Below
Product Details & Documents
This VWR® glove with polyurethane coating provides superior cut protection for applications that include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coating on palm and fingertips
  • Grey knitted glove with grey coating
Ideal for handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for EN 388:2016 (4X42E). Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-5691
  • 112-5692
  • 112-5693
  • 112-5694
  • 112-5695
  • 112-5696
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Finish:
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42E)
  • EN 388:2016 (4X42E)
  • EN 388:2016 (4X42E)
  • EN 388:2016 (4X42E)
  • EN 388:2016 (4X42E)
  • EN 388:2016 (4X42E)
Specifications
Product Family Options
Product Information
  • 手袋のサイズ
    Colour
    Availability
    Price
  • 6
    Grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5691
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X42E)
  • 7
    Grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5692
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X42E)
  • 8
    Grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5693
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X42E)
  • 9
    Grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5694
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X42E)
  • 10
    Grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5695
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X42E)
  • 11
    Grey
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5696
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    13
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X42E)