Orders LinkContactUsLinkComponent
マイクロ遠心機チューブ、 クラシック
マイクロ遠心機チューブ、 クラシック
マイクロ遠心機チューブ、 クラシック
Supplier:  VWR Collection
Certificates
マイクロ遠心機チューブ、 クラシック
マイクロ遠心機チューブ、 クラシック
15 Options Available Below
Product Details & Documents
VWR® microcentrifuge tubes are manufactured using FDA approved resin and these are free from RNase, DNase, DNA, pyrogens and trace metals.
  • Can be centrifuged up to 20000 RCF
  • Withstands temperatures from –80 to +121 °C
  • Easy-seal cap design for smooth single hand operations
  • Transparent surface for hindrance-free spectrophotometer reading
  • Thin cap membranes in tubes for trouble free needle piercing
  • Flat, frosted lid and larger surface on body for easy marking and labelling
Crafted with precision, these top-tier tubes excel in delivering unparalleled performance throughout storage, isolation, centrifugation, and sample preparation procedures. They stand as the preferred option across diverse disciplines such as microbiology, molecular biology, biochemistry, and an extensive spectrum of bioanalytical techniques.
The 0,5, 1,7 and 2 ml tubes are made in a Class 100000 cleanroon facility to affirm particulate-free and tracemetal-free products.
Product Family Options
Product Information
  • 容量
    Colour
    Type
    Version
    RCF max.
    Packed
    Availability
    Price
  • 0,5 ml
    Natural
    Conical bottom, flat cap
    Non sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $14.50
    $14.50
    1 * 1.000 items
       $14.50
  • 0,5 ml
    Natural
    Conical bottom, flat cap
    Sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $18.32
    $18.32
    1 * 1.000 items
       $18.32
  • 0,5 ml
    Amber
    Conical bottom, flat cap
    Non sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $175.45
    $175.45
    1 * 1.000 items
       $175.45
  • 0,5 ml
    Amber
    Conical bottom, flat cap
    Sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $213.59
    $213.59
    1 * 1.000 items
       $213.59
  • 0,5 ml
    Natural
    Conical bottom, flat cap
    Sterile, LR
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $309.02
    $309.02
    1 * 1.000 items
       $309.02
  • 1,7 ml
    Natural
    Conical bottom, flat cap
    Non sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $155.68
    $155.68
    1 * 1.000 items
       $155.68
  • 1,7 ml
    Natural
    Conical bottom, flat cap
    Sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $190.71
    $190.71
    1 * 1.000 items
       $190.71
  • 1,7 ml
    Amber
    Conical bottom, flat cap
    Non sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $172.91
    $172.91
    1 * 1.000 items
       $172.91
  • 1,7 ml
    Amber
    Conical bottom, flat cap
    Sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $211.05
    $211.05
    1 * 1.000 items
       $211.05
  • 1,7 ml
    Natural
    Conical bottom, flat cap
    Sterile, LR
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $419.56
    $419.56
    1 * 1.000 items
       $419.56
  • 2,0 ml
    Natural
    Conical bottom, flat cap
    Non sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $104.39
    $104.39
    1 * 500 items
       $104.39
  • 2,0 ml
    Natural
    Conical bottom, flat cap
    Sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $120.78
    $120.78
    1 * 500 items
       $120.78
  • 2,0 ml
    Amber
    Conical bottom, flat cap
    Non sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $127.14
    $127.14
    1 * 500 items
       $127.14
  • 2,0 ml
    Amber
    Conical bottom, flat cap
    Sterile
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $165.28
    $165.28
    1 * 500 items
       $165.28
  • 2,0 ml
    Natural
    Conical bottom, flat cap
    Sterile, LR
    20000×g
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $24.80
    $24.80
    1 * 500 items
       $24.80