Orders LinkContactUsLinkComponent
デジタルディスクローテーター, VWR®
Digital Disc Rotator
Catalog # 521-2779
Supplier:   VWR Collection
デジタルディスクローテーター, VWR®
Catalog # 521-2779
Supplier:  VWR Collection
デジタルディスクローテーター, VWR®
デジタルディスクローテーター, VWR®
Digital Disc Rotator
Product Details & Documents
The Digital disc rotator is the result of extensive research and user experience feedback. Consisting of a maintenance free brushless DC motor, the rotator offers powerful performance without any hassel. The features such as intermittent mode, user programmable setup and its pulse mode offer thorough mixing without much effort.
  • Microprocessor controlled disc rotator for intelligent performance with last run memory feature
  • Variable angle setting to adjust the rotator angle from 0 to 90°
  • User programmable mode to set 9 user-defined programs for greater convenience
  • Auto power on feature starts the unit automatically after a power failure and runs for the remaining time left
  • Safe low voltage device
  • Multiple attachments – 12×50 ml/24×15 ml/44×2 ml tubes available as optional accessories
Certifications: IP protection class according to DIN EN 60529: IP 21
Ordering information: Disc rotator 521-2779 is supplied without the disc plate which must be ordered separately.
Specifications
  • 説明:
    Digital Disc Rotator
  • Seq:
    01
  • Cat. No.:
    521-2779
  • Tilt angle:
    0 - 90°
  • Speed:
    10 - 80 min⁻¹
  • Timer:
    1 - 99 min and infinite mode
  • W×D×H:
    383×325×325 mm
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • VWRA444-1166
    Disk for 12×50 ml tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • VWRA444-1170
    Disk for 44×2ml tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • VWRA444-1167
    50 ml tube clamp
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • VWRA444-1168
    Disk for 24×15 ml tubes
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • VWRA444-1169
    15 ml tube clamp
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • VWRA444-1171
    5/2 ml tube clamp