臨床用遠心分離機、VWR® (ブイアール
Clinical centrifuge
Catalog # VWRA521-2803
Supplier: VWR Collection
Jump to:
Product Details & Documents
This clinical centrifuge is equipped with quality components and features for a superior user expereince. With multiple rotor options, it can accommodate a wide range of sample sizes which makes it extremely versatile. The device has also been designed with a compact footprint to occupy less space relative to its functionality.
- Microprocessor controlled for efficient performance even under heavy loads
- Brushless DC motor for maintenance free long life
- Favorite mode allows user to set up to 99 user defined programs
- Enhanced airflow design to keep the temperature under control
- Lid lock safety feature – lid opens automatically when is run completed
- One touch RPM / RCF conversion and short spin operation option on keypad
- Safe low voltage device
注文情報: Supplied without rotors. A range of additional optional rotors are available and must be ordered separately.
納品情報: Supplied with reduction adaptors for various sizes of blood collection tubes: 9, 6, 4, 3 and 2 ml tubes.
Documents
VWR® Centrifuges
Product Literature
Specifications
- 説明:Clinical centrifuge
- Cat. No.:VWRA521-2803
- Timer:1 - 999 min and infinity mode
- Max. speed:4000 min⁻¹
- Dimensions:415×355×173 mm
- Max. RCF:2270 ×g
- Weight:7,7 kg
- Display:Digital