VWR®, Cut Resistant Gloves, PU Coating
Supplier: VWR Collection
Certificates
About this item
This VWR® glove with polyurethane coating provides general cut protection for the widest range of applications.
- ANSI A2 Cut Protection (Level B)
- UHMWPE/nylon/spandex composition
- Polyurethane (PU) coating on palm and fingertips
- Grey knitted glove with grey coating
Ideal for handling items or tools with a risk of cuts, including material handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A2 Cut Resistance. EN 388:2016 (4X42B)
Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications
- Catalog No:
- 112-5676
- 112-5678
- 112-5677
- 112-5673
- 112-5675
- 112-5674
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Finish:
- PU palm coating
- PU palm coating
- PU palm coating
- PU palm coating
- PU palm coating
- PU palm coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
Specifications
Related Information
VWR® Cut Protection Gloves, PU Coating
Additional Product Details
VWR® Reusable Cut Resistant and Precision Work Glove Guide