Orders LinkContactUsLinkComponent
プラットフォームシェーカー、VWR
プラットフォームシェーカー、VWR
プラットフォームシェーカー、VWR
プラットフォームシェーカー、VWR
These platform shakers combine performance with user-friendliness for enhanced convenience. With a powerful brushless DC motor and top-of-the line components, units are fully equipped to deliver exceptional performance. Leading features like loop function and user programmable interface ensure the device is convenient and easy to use.
  • Microprocessor controlled for efficient performance even under high loads
  • Variable speed selection up to 500 min⁻¹ (10 mm) and up to 300 min⁻¹ (20 mm)
  • Optional RS232 interface for remote operation
  • Auto power on in case of power failure
  • Choice of models with orbital or linear motion
 : IP Protection Class according to DIN EN 60259: IP 21.
 : Supplied without accessories. Attachments must be ordered separately.
  • Catalog No:
  • VWRA444-1229
  • VWRA444-1230
  • Speed:
  • 50 - 500 min⁻¹
  • 50 - 500 min⁻¹
  • Shaking motion:
  • Orbital
  • Linear
  • Max. load:
  • 10 kg
  • 10 kg
  • Weight:
  • 7,5 kg
  • 7,5 kg
  • W×D×H:
  • 350×408×99 mm
  • 350×408×99 mm
Product Family Options
Platform orbital shaker 10 mm
  • 説明
  • Orbital platform shaker
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    VWRA444-1229
    Speed
    50 - 500 min⁻¹
    Shaking motion
    Orbital
    Max. load
    10 kg
    Weight
    7,5 kg
    W×D×H
    350×408×99 mm
Platform linear shaker 10 mm
  • 説明
  • Linear platform shaker
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    VWRA444-1230
    Speed
    50 - 500 min⁻¹
    Shaking motion
    Linear
    Max. load
    10 kg
    Weight
    7,5 kg
    W×D×H
    350×408×99 mm
Accessories
  •  
    説明
  • VWRA444-1233
    Universal platform with adjustable bars (flask capacity: 13×100 ml; 13×150ml; 9×250 ml; 6×500 ml or 5×1000 ml)
    1 * 1 items
  •  
    説明
  • VWRA444-1234
    Disk attachment base plate with slip resistant tray
    1 * 1 items
  •  
    説明
  • VWRA444-1235
    Funnel attachment
    1 * 1 items
  •  
    説明
  • VWRA444-1236
    Base plate for plastic clamps (clamp capacity: 9×250 ml or 9×500 ml)
    1 * 1 items
  •  
    説明
  • VWRA444-1237
    Plastic clamp for 250 ml flasks
    1 * 1 items
  •  
    説明
  • VWRA444-1238
    Plastic clamp for 500 ml flasks
    1 * 1 items
  •  
    説明
  • VWRA444-1239
    Base plate for stainless steel clamps (clamp capacity: 9×100 ml; 9×150 ml or 9×250 ml)
    1 * 1 items
  •  
    説明
  • VWRA444-1240
    Stainless steel clamp for 100 ml flasks, suitable for base plate
    1 * 1 items
  •  
    説明
  • VWRA444-1241
    Stainless steel clamp for 150 ml flasks, suitable for base plate
    1 * 1 items
  •  
    説明
  • VWRA444-1242
    Stainless steel clamp for 250 ml flasks, suitable for base plate
    1 * 1 items
  •  
    説明
  • VWRA444-1243
    Base plate for stainless steel clamps (clamp capacity: 9×500 ml or 5×1000 ml)
    1 * 1 items
  •  
    説明
  • VWRA444-1244
    Stainless steel clamp for 500 ml flasks, suitable for base plate
    1 * 1 items
  •  
    説明
  • VWRA444-1245
    Stainless steel clamp for 1000 ml flasks, suitable for base plate
    1 * 1 items