Orders LinkContactUsLinkComponent
VWR®遠心分離機用チューブ、ASTM
VWR®遠心分離機用チューブ、ASTM
Supplier:  VWR Collection
Certificates
VWR®遠心分離機用チューブ、ASTM
VWR®遠心分離機用チューブ、ASTM
2 Options Available Below
Product Details & Documents
Borosilicate glass 3.3, used for the determination of water and sediments in crude oils according to test method ASTM D 96.
  • High chemical and thermal resistance
  • High contrast blue enamel graduations
  • Withstand forces up to 700 ×g
Design, graduations and tolerances meet ASTM (American Society for Testing Materials) specifications.
Certifications: ASTM E-982
Delivery information: Delivered without cap.
Product Family Options
Product Information
  • 説明
    容量
    直径×長さ
    Closure type
    Availability
    Price
  • Centrifuge tube, conical
    100 ml
    17×200 mm
    Without cap
    1 * 1 items
  • Centrifuge tube, pear-shaped
    100 ml
    17×200 mm
    Without cap