VWR®, ラボ乾燥ラック
Drying rack set, 441×660×139 mm
Catalog # 149-0297
Supplier: VWR Collection
Jump to:
Product Details & Documents
Versatile rack made of durable, easy to clean PS.
- Channels designed for more useful configurations
- Use on counter or mount on wall
- Large and small pegs for all glassware varieties
- One of a kind drying basket to collect small items
Designed to protect against breakage
- Peg angle allows for better hold - especially for large, heavy pieces
- Patent pending SureGrip hook securely grips glassware into place
- Holds up to 35 kg
Delivery information: Includes 72 pegs (36 small, green and 36 large, blue), 2 drying baskets (clear), drainage tubing, mounting hardware.
Specifications
- 説明:Drying rack set
- W×D×H:441×660×139 mm