

Magnesium perchlorate, AR® ACS desiccant, Macron Fine Chemicals™
MACR5039-04
:
- パックタイプ:Glass bottle, amber, wide mouth
- Pk:500 g
- Seq:3645
- Pk Info:1×500 g
- Loss on Drying at 190°C:≤8 %
- ACS - Suitability for Moisture Absorption:Passes Test
- ACS - Titrable Free Acid (meq/g):≤0.005
- ACS - Titrable Base (meq/g):≤0.025
:
:223.21 g/mol
:13,05702
:251 °C
:2,60 g/cm³
:233-108-3
:1475
:5.1,II
:Management of Change (MOC) category = N
:GHS07
:GHS03
:10034-81-8
:Cl₂MgO₈
:MFCD00011102