Dextrose kit, anhydrous USP
Kit, 1× 5 kg + 1× 350 g, 1 KIT
Catalog # 1919-X2
Supplier: Avantor
Jump to:
Product Details & Documents
Synonyms: Corn sugar, Glucose, D-(+)-Dextrose, Glucolin, Dextrose, D-Glucose
- CAS Number:50-99-7
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00148912
- Molecular Weight:180.16 g/mol
- Merck Index:14,04459
- Melting Pt:146 °C
- Density:1,54 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-075-1
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specifications
- パックタイプ:Kit
- Pk:1 KIT
- Pk Info:1× 5 kg + 1× 350 g



