Orders LinkContactUsLinkComponent
L字型セルスプレッダー、無菌
L字型セルスプレッダー、無菌
Supplier:   VWR Collection
L字型セルスプレッダー、無菌
Supplier:  VWR Collection
Certificates
L字型セルスプレッダー、無菌
L字型セルスプレッダー、無菌
2 Options Available Below
Product Details & Documents
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
  • L-shape allows application on the entire dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damage to samples
  • Sterilised by electron beam irradiation
  • Non-pyrogenic
Spreaders made of polypropylene and pre-sterilised, eliminating the need for flaming or autoclaving.
Certifications: Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
包装: Individual wrap and bulk packaging available.
Product Family Options
Product Information
  • Colour
    Packed
    Availability
    Price
  • Green
    Individually packed
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $10.46
    $10.46
    1 * 100 items
       $10.46
  • Green
    Bulk packed; 10/bag
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $28.83
    $28.83
    1 * 500 items
       $28.83