VWR® DuPont™ Tyvek® IsoClean® Material overalls, with hood
VWRU89012-818
- 衣服のサイズ:XXL
- Anti-static:+
- Garment application:Laboratory
- Colour:White
- 説明:Hooded coveralls with elastic ankles
- Pk:25
Overalls provide high levels of protection and durability while remaining comfortable. A unique flash spinning process creates an excellent barrier against dry particulates, non-hazardous liquids, and microorganisms.
- Protective, durable, and comfortable
- Excellent barrier against dry particulates, non-hazardous liquids, and microorganisms
- Chemically and biologically inert, low linting, and static free until laundered
- Serged seams, front zipper closure, elastic wrists, and attached hood
- Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles
Antistatic overalls are low-linting and chemically and biologically inert.