Orders LinkContactUsLinkComponent
VWR®, 試薬ボトル、ワイドネック
Supplier:   VWR Collection
VWR®, 試薬ボトル、ワイドネック
Supplier:  VWR Collection
Certificates
VWR®, 試薬ボトル、ワイドネック
VWR®, 試薬ボトル、ワイドネック
13 Options Available Below
Product Details & Documents
Made from clear or amber soda-lime glass. Ideal for sampling, storage of powders or liquids, or other applications that do not involve high thermal stress.
  • Good chemical and physical properties
  • Clear or amber glass, available with or without black PP screw cap
  • Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
 : Thread in accordance with DIN 168.
Product Family Options
Clear glass without cap
Product Information
  • 容量
    Thread
    外径
    高さ
    Colour
    Availability
    Price
  • 30 ml
    GL 32
    36 mm
    68 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 50 ml
    GL 32
    44 mm
    75 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 85 items
    1 * 85 items
  • 50 ml
    GL 40
    44 mm
    69,5 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 125 items
    1 * 125 items
  • 100 ml
    GL 40
    50 mm
    92,5 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 63 items
    1 * 63 items
  • 125 ml
    GL 40
    54 mm
    98 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 80 items
    1 * 80 items
  • 150 ml
    GL 45
    57 mm
    104 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 42 items
    1 * 42 items
  • 200 ml
    GL 55
    63 mm
    109 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 56 items
    1 * 56 items
  • 250 ml
    GL 55
    69 mm
    113 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 48 items
    1 * 48 items
  • 500 ml
    GL 55
    84 mm
    154 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 20 items
    1 * 20 items
  • 1000 ml
    GL 68
    103,2 mm
    180,5 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 18 items
    1 * 18 items
Amber glass with cap
Product Information
  • 容量
    Thread
    外径
    高さ
    Colour
    Availability
    Price
  • 30 ml
    GL 32
    36 mm
    68 mm
    Amber
    Log in for availability
    Log in for availability
    1 * 126 items
    1 * 126 items
  • 50 ml
    GL 32
    44 mm
    75 mm
    Amber
    Log in for availability
    Log in for availability
    1 * 85 items
    1 * 85 items
  • 100 ml
    GL 40
    50 mm
    92,5 mm
    Amber
    Log in for availability
    Log in for availability
    1 * 63 items
    1 * 63 items