Order Entry
Japan
ContactUsLinkComponent
 
Irritant
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99% (dried basis), purified, J.T.Baker®
Catalog # 4910-09
Supplier:  Avantor
CAS Number:  
Irritant
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99% (dried basis), purified, J.T.Baker®
Catalog # 4910-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

Specification Test Results

  • Assay (dried basis):
    ≥99 %
  • Appearance:
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • pH of 1% Solution at 25°C:
    2.5 - 4.5
  • Solution (10%) in Water:
    Passes Test
  • Loss on Drying:
    ≤1 %
  • Absorbance of a 0.1M Solution (1-cm path vs water) at - (1 cm path vs water at 260 nm):
    ≤0.020
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Iron (Fe):
    ≤5 ppm
  • Equivalent Weight, gmol:
    207.2 - 211.4
  • Optical Density:
    ≤0.1
  • Water (by KF titration):
    ≤0.1 %

Specification Test Results

About this item

 :
 :209.27 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1132-61-2
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183