
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, FCC, J.T.Baker®
Catalog # 3600-B1
Supplier: Avantor
CAS Number:
Specifications
- Pk:13,748 kg
- Seq:4205
Specifications
Specification Test Results
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Water (H₂O):12.0 - 15.0 %
- NF - Assay (Na₂CO₃) (anhydrous basis):99.5 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤4 mg/kg
- FCC - Loss on Drying:12.0 - 15.0 %
- FCC - Assay (Na₂CO₃) (anhydrous basis):99.5 - 100.5 %
Specification Test Results
About this item
:
:124.00 g/mol
Density:2,25 g/cm³ (20 °C)
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient
CAS Number:5968-11-6
Formula:Na₂CO₃·1H₂O
MDL Number:MFCD00149177