Orders LinkContactUsLinkComponent
VWR®, アクセサリー
Supplier:   VWR Collection
VWR®, アクセサリー
Supplier:  VWR Collection
Certificates
VWR®, アクセサリー
VWR®, アクセサリー
8 Options Available Below
Product Family Options
Product Information
  • 説明
    Availability
    Price
  • Sipper unit with temp. control, A-1000; controller (constant-temperature Peltier sipper system A1000, used for UV1600PC, UV-3100PC, V-3000PC, UV6300PC; cell no.: 1; path length: 10 mm; temp. range: 10 ~ 90 °C [0,1 °C interval adjustable]; temp. control accuracy: ±0,1 °C; injection speed: 1 ~ 8 ml/min [0,5 ml/min interval adjustable])
    1 * 1 items
  • Peltier unit, A-1200; controller, constant-temperature system A1200, used for UV1600PC, UV-3100PC, V3000PC, UV6300PC, cell no. 1, path length: 10 mm, temp. range: 10 ~ 90 °C [0,1 °C interval adjustable (temp. control accuracy : ±0,1 °C)]
    1 * 1 items
  • Controller, sipper system A1100, sipper unit without peltier temp. control (used for UV1600PC, UV-3100PC, V3000PC, UV6300PC; cell no.: 1, path length: 10 mm; injection speed : 1 ~ 8 ml/min. [0,5 ml/min interval adjustable])
    1 * 1 items
  • Deuterium lamp
    1 * 1 items
  • M. wave professional software
    1 * 1 items
  • Replacement battery, 3,6 V, CR2032
    1 * 1 items
  • Thermal paper for printer, 57 mm, 50 m, Core ¹/₂"
    Log in for availability
    Log in for availability
    1 * 4 Roll
    1 * 4 Roll
  • Thermal printer T58KP, ROHS certified