Orders LinkContactUsLinkComponent
VWR®, 試薬リザーバー
VWR®, 試薬リザーバー
VWR®, 試薬リザーバー
VWR®, 試薬リザーバー
Designed for use in multi-channel pipetting applications, these disposable reservoirs accommodate all types of pipettes up to 12-channel width.
  • Trough design, graduation marks on interior walls, and pour-off spouts on each corner
  • Lot certified, non pyrogenic, DNase- and RNase-free
  • Non sterile or electron beam sterilised
 : 25 ml reservoirs also available in a unique divided format that features three distinct mini reservoir chambers, each with a 5 ml capacity; 50 ml reservoirs available in clear FDA-grade PVC or virgin-modified PS; all other sizes manufactured from virgin-modified PS.
Product Family Options
25 ml
  • Type
    容量
    Sterile
    Packed
  • PS
    25 ml
    -
    50/case
    1 * 50 items
    1 * 50 items
  • PS, 3 divisions
    25 ml
    -
    100/case
    1 * 100 items
    1 * 100 items
  • PS, 3 divisions
    25 ml
    +
    5/bag, 40 bags/case
    1 * 200 items
    1 * 200 items
  • PS, 3 divisions
    25 ml
    +
    1/bag, 100/case
    1 * 100 items
    1 * 100 items
50 ml
  • Type
    容量
    Sterile
    Packed
  • PVC
    50 ml
    +
    1/bag, 100/case
    1 * 100 items
    1 * 100 items
  • PVC
    50 ml
    +
    5/bag, 40 bags/case
    1 * 200 items
    1 * 200 items