Orders LinkContactUsLinkComponent
VWR®, ゲルローディング用ピペットチップ
VWR®, ゲルローディング用ピペットチップ
VWR®, ゲルローディング用ピペットチップ
Supplier:  VWR Collection
Certificates
VWR®, ゲルローディング用ピペットチップ
VWR®, ゲルローディング用ピペットチップ
4 Options Available Below
Product Details & Documents
VWR® gel-loading pipette tips faciliate precision sample loading into gels.
  • Standard 96 tip format is multi-channel friendly
  • Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available)
  • Sterile product is also pyrogen-free
Universal fit to accommodate most pipettes except Rainin LTS and Thermo Clip-Tip.
Ordering information: Choose from sterile or non sterile formats, racked or bulk packaging.
Product Family Options
Product Information
  • Tip style
    容量
    Sterile
    長さ
    Packed
    Availability
    Price
  • Round tip, Ø 0,57 mm
    200 µl
    -
    83 mm
    6 racks of 96
    Log in for availability
    Log in for availability
    1 * 576 items
    1 * 576 items
  • Round tip, Ø 0,57 mm
    200 µl
    +
    83 mm
    6 racks of 96
    Log in for availability
    Log in for availability
    1 * 576 items
    1 * 576 items
  • Round tip, Ø 0,57 mm
    200 µl
    -
    83 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Round tip, Ø 0,57 mm
    200 µl
    -
    83 mm
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $149.25
    $149.25
    1 * 1.000 items
       $149.25