VWR®, Digital Hotplate Stirrers, 350 and 550 °C
Discover to do more these two temperature range hotplate stirrers. They have a leading feature which saves time as well as effort, allowing to accomplish more with less effort. They have three heating modes.
- Robust top made up of nanocrystalline glass ceramic for resistance against temperature shocks and liquid splash
- Elevated control panel for protection against leaking liquids
- Display for easy value setting, all set values readily visible without toggle
- No shared display
- Heating range from ambient to 350/550 °C
- Intelligent over-temperature control switches off the Hotplate
- Variable speed setting from 200 to 2200 min⁻¹ in steps of 10 min⁻¹
- Safe temperature setting up to 375/575 °C for protection against overheating and better reliability
Timer Range: 1 to 999 minutes and infinite mode.
Slip detection mode for detecting stir bar slip and automatic optimization of stirring operations to ideal limit.
: IP Protection Class: IP 21.
: Accessories, stand and PT-1000 probe to be ordered seperately.
- Catalog No:
- VWRA444-1206
- VWRA444-1205
- Speed range:
- 2200 min⁻¹
- 2200 min⁻¹
- Max. stirring capacity H₂O:
- 20 L
- 20 L
Frequently Bought Together