スターラー、2/4/6 L容量、VWR®
Supplier: VWR Collection
Certificates
About this item
Digital hot plate stirrers with 5, 10, and 15 stirring positions are engineered to offer unparalleled efficiency. Both the software and hardware of these devices are tailored to maximize productivity and safety with their exceptional performance. They come equipped with a programmable pulse and a heating efficiency mode for an intuitive user experience.
- High performance multi position stirrer with 5, 10 and 15 stirring positions with optimum use of laboratory space
- Variable speed ranging from 300 to 1500 min⁻¹ in steps of 10 min⁻¹
- Timer setting from 1 to 999 min and infinite
- Heater temperature setting from ambient to 120 °C
- Homogeneous temperature distribution across heating plate
- Closed design allows easy cleaning and protects the equipments from accidental spills
- Water proof heater with inbuilt safety cutoff
- Non slip strength rubber feet
Program will remain the same for all positions.
Plate material: Stainless steel plate with silicone sheet (setup plate)
Certifications: IP Protection Class: IP42
Ordering information: Along with main unit, standard Stir bar 25 mm are included.
Specifications
- Catalog No:
- VWRA444-1212
- VWRA444-1213
- VWRA444-1214
- Model:
- VWRA444-1212
- VWRA444-1213
- VWRA444-1214
- Plate dimensions:
- 565×136×75 mm
- 565×196×75 mm
- 565×282×75 mm
- No. of stirring positions:
- 5
- 10
- 15
- Speed range:
- 300 - 1500 min⁻¹
- 300 - 1500 min⁻¹
- 300 - 1500 min⁻¹
- Timer:
- 1 - 999 min. and infinite (run time)
- 1 - 999 min. and infinite (run time)
- 1 - 999 min. and infinite (run time)
- Temperature range:
- Ambient…120 °C
- Ambient…120 °C
- Ambient…120 °C