Orders LinkContactUsLinkComponent
VWR® PHPピペットスタンド
VWR® PHPピペットスタンド
Supplier:   VWR Collection
VWR® PHPピペットスタンド
Supplier:  VWR Collection
Certificates
VWR® PHPピペットスタンド
VWR® PHPピペットスタンド
2 Options Available Below
Product Details & Documents
The VWR® Pipette Stand is a rotatable stand that conveniently holds up to six single and multi-channel pipettes. This helps users to maintain not only a tidy lab environment, but it also reduces the risk of pipette damage or cross-contamination.
  • Rotating (carousel) stand structure
  • Can hold up to six pipettes
  • Robust design
  • Efficient use of space
Product Family Options
Product Information
  • 説明
    Availability
    Price
  • PHP Carousel stand for 6 pipettes
    1 * 1 items
  • PHP Shelf mounting stand for Pipette