Orders LinkContactUsLinkComponent
クリーンルーム用フェイスマスク、クラス100
クリーンルーム用フェイスマスク、クラス100
クリーンルーム用フェイスマスク、クラス100
クリーンルーム用フェイスマスク、クラス100
Cleanroom face masks are manufactured with non woven polypropylene/polyethylene (ES) / melt-blown polypropylene (MBPP) materials.
  • Low particle generation - suitable for critical environments
  • High bacterial filtration efficiency and low breathable resistance
  • Packed in a cleanroom environment
  • Solvent-safe double-bag cleanroom packaging
  • 30% nylon/70% polyester ear loops
  • Plastic wire nose clip
  • For industrial and cleanroom use only; do not use for protection against COVID-19
  • This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745 / Directive 93/42/EEC
  • This product is not a personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425
These face masks are designed for applications in critical environments, such as semiconductor, electronic industries, or high-tech industries. Masks should be stored at room temperature, in a dry and shaded environment. Recommended shelf life is 3 years from the date of manufacture.
Bacterial filtration efficiency: >95%
Delta P: <6 mm H₂O
 : 100 masks per double-bag, 20 bags per case.
Product Family Options
  • Type
    サイズ
    Colour
  • with ear loops
    17,5 L× 9,5 W cm
    White
    1 * 100 items
    1 * 100 items
  • with headband
    21 L × 9,5 W cm