Orders LinkContactUsLinkComponent
VWR®, ガス密閉式マイクロシリンジ
Supplier:   VWR Collection
VWR®, ガス密閉式マイクロシリンジ
Supplier:  VWR Collection
Certificates
VWR®, ガス密閉式マイクロシリンジ
VWR®, ガス密閉式マイクロシリンジ
9 Options Available Below
Product Details & Documents
Micro syringes without needle
General purpose Luer lock connector for use with autosamplers and liquid handling instruments. All parts in contact with sample are made from neutral borosilicate glass and PTFE. This provides excellent performance and an inert sample environment.
Interchangeable needles
549-0560 made from SUS304; 549-0565 made form chrome plated brass. 549-0560 needles for 1; 2,5; 5 and 10 ml gas-tight syringes. 549-0565 needles for 0,1 ml gas-tight syringes.
PTFE tips
Provides a complete leak-tight seal for both liquids and gases.
549-0560 needles for 1, 2,5, 5 and 10 ml gas-tight syringes. 549-0565 needles for 0,1 ml gas-tight syringes.
 : Needles must be ordered separately.
Product Family Options
Micro syringes without needle
Product Information
  • 説明
    容量
    長さ
    Type of tip
    For
    Availability
    Price
  • Gas-tight micro syringe
    1 ml
    1 * 1 items
  • Gas-tight micro syringe
    2,5 ml
    1 * 1 items
  • Gas-tight micro syringe
    5 ml
    1 * 1 items
  • Gas-tight micro syringe
    10 ml
    1 * 1 items
Micro syringe with interchangeable needle
Product Information
  • 説明
    容量
    長さ
    Type of tip
    For
    Availability
    Price
  • Gas-tight micro syringe
    0,10 ml
    50 mm
    Bevelled
    1 * 1 items
PTFE tips
Product Information
  • 説明
    容量
    長さ
    Type of tip
    For
    Availability
    Price
  • PTFE tips
    1 ml micro syringe
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • PTFE tips
    2,5 ml micro syringe
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • PTFE tips
    5 ml micro syringe
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • PTFE tips
    10 ml micro syringe
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
Interchangeable needles
Product Information
  • Catalog #
    Type of tip
    外径
    Gauge
    説明
    内径
    長さ
    Availability
    Price
  • Bevelled
    0,52 mm
    -
    Interchangeable without PTFE seal
    0,18 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - $0.00
    $0.00
    1 * 5 items
       $0.00
Product Information
  • Catalog #
    Type of tip
    外径
    Gauge
    説明
    内径
    長さ
    Availability
    Price
  • Bevelled
    0,56 mm
    24
    Luer lock with PTFE seal
    0,30 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - $0.00
    $0.00
    1 * 5 items