VWR® ARISTAR® Multi-Component Anion Mix 9, Avantor
Catalog # 458164C
Supplier: VWR Chemicals
CAS Number:
Specifications
- パックタイプ:Plastic bottle
- Pk:1 SET
- 説明:Multi-Component Anion Mix 9
Specifications
About this item
Two solutions - a nitrite standard solution and a 6 component multi-element standard solution.
Br 40 ppm NO3 40 ppm
Cl 40 ppm PO4 40 ppm
F 20 ppm SO4 40 ppm
NO2 40 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.