VWR® ARISTAR® Anion Multi-Component Standard 1 for Ion Chromatography, Avantor
Catalog # 458142C
Supplier: VWR Chemicals
CAS Number:
Specifications
- パックタイプ:Plastic bottle
- Pk:100 mL
- 説明:Anion multi-component standard 1 for IC
Specifications
About this item
Cl 30 ppm PO4 150 ppm
F 20 ppm SO4 150 ppm
NO3 100 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.