VWR® ARISTAR®, ICP Multi-Element Calibration Standard 1 in dilute Nitric Acid
Catalog # 456452F
Supplier: VWR Chemicals
CAS Number:
Specifications
- パックタイプ:Plastic bottle
- Pk:100 mL
- 説明:Multi-element calibration standard 1 in dil. nitric acid ARISTAR® for ICP
Specifications
About this item
In 5% nitric acid solution.
19 Elements : 50 mg/L Ag, Ni; 100 mg/L Al; 5 mg/L B, Fe 15 mg/L, Ba, Mn; 1 mg/L Be, Sr; 200 mg/L Bi, In, Pb; 20 mg/L Cd, Co, Cu, Zn; 25 mg/L Cr; 150 mg/L Ga; 40 mg/L Tl
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.