pH Indicator solution, BDH® '4.5' pH 3,5 - 6,0 for titration of first hydrogen of phosphoric acid
Supplier: VWR Chemicals
Certificates
About this item
:
Boiling Pt:82,8 °C (1013 hPa)
Flash Pt:18 °C (closed cup)
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS02
:GHS07
Related Information
Certificates
Search by Lot Numberspec_VWRC210414M
Product specification
VWR® Indicators