Orders LinkContactUsLinkComponent
VWR®, マイクロシリンジ超微量
VWR®, マイクロシリンジ超微量
Supplier:   VWR Collection
VWR®, マイクロシリンジ超微量
Supplier:  VWR Collection
Certificates
VWR®, マイクロシリンジ超微量
VWR®, マイクロシリンジ超微量
2 Options Available Below
Product Details & Documents
Micro syringe with fixed needle for high sensitivity GC analysis and ultra-minute volumes.
  • Zero dead volume between plunger and needle
  • Fixed needle with bevelled tip for smooth septum penetration
Length in ordering table refers to needle length.
Product Family Options
Product Information
  • 容量
    長さ
    Type of tip
    Availability
    Price
  • 5 µl
    50 mm
    Bevelled
    1 * 1 items
  • 1 µl
    50 mm
    Bevelled