VWR®, 相分離紙、グレード511
Supplier: VWR Collection
About this item
Hydrophobic filter paper used for separation of aqueous solutions from organic solutions.
Usable as a disposable separating funnel
Paper is impregnated with a special silicon coating which makes the paper water-repellent for facilitating the separation of water and solvents.
5 Options Available Below
Product Details & Documents
Hydrophobic filter paper used for separation of aqueous solutions from organic solutions.
Usable as a disposable separating funnel
Paper is impregnated with a special silicon coating which makes the paper water-repellent for facilitating the separation of water and solvents.
Specifications for Product Family
Specifications
- Catalog No:
- 516-0313
- 516-0314
- 516-0315
- 516-0311
- 516-0312
- Weight:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Thickness:
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- 0,2 mm
- Filtration speed:
- 200 s (Herzberg)
- 200 s (Herzberg)
- 200 s (Herzberg)
- 200 s (Herzberg)
- 200 s (Herzberg)
Specifications
Product Family Options
Product Information
- GradeサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.516-0313Weight85 g/m²Thickness0,2 mmFiltration speed200 s (Herzberg) - Specifications:
More
Specifications
Cat. No.516-0314Weight85 g/m²Thickness0,2 mmFiltration speed200 s (Herzberg) - Specifications:
More
Specifications
Cat. No.516-0315Weight85 g/m²Thickness0,2 mmFiltration speed200 s (Herzberg) - Specifications:
More
Specifications
Cat. No.516-0311Weight85 g/m²Thickness0,2 mmFiltration speed200 s (Herzberg) - Specifications:
More
Specifications
Cat. No.516-0312Weight85 g/m²Thickness0,2 mmFiltration speed200 s (Herzberg)