Orders LinkContactUsLinkComponent
VWR®, 抽出用シンブル、グレード501
VWR®, 抽出用シンブル、グレード501
Supplier:  VWR Collection
Certificates
VWR®, 抽出用シンブル、グレード501
VWR®, 抽出用シンブル、グレード501
12 Options Available Below
Product Details & Documents
Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
  • Determination of free lipids content in foods and fat content in milk powder
  • Determination of oil content in oleaginous products
  • Extraction and quantification of various components
Product Family Options
Product Information
  • Int.Ø×H
    Availability
    Price
  • 19×90 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 22×80 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 25×80 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 25×100 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 28×80 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 30×80 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 30×100 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 33×60 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 33×80 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 33×100 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 33×118 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • 41×123 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items