VWR® Universal Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
- Compatible with a wide range of pipettes
- Maximizes sample recovery with low binding resin and a fine point design
- Improves ergonomics by reducing the force required to attach and eject a tip
- Autoclavable at 15 psi, 122 °C for 15 minutes
- Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
- Type de test:Sandwich
- Méthode de détection:Sandwich ELISA, double antibody
- Primary antibody reactivity:Human
- Portée de détection:7,813 - 500 pg/ml
- Type déchantillon:Serum, plasma, cell culture supernatant, cell or tissue lysate, other liquid samples
- Time to Results:4 hours
- Température de stockage:Refrigerate (2…8 °C)
- Cat. No.:GENOIT2386
- Format:Pre-coated
- Cible:IL-15RA
- Sensibilité:4,688 pg/ml
- Regulatory status:RUO
- Durée de conservation:12 Months