Orders LinkContactUsLinkComponent
VWR®, 臨床用4ウェイチューブラック、
Supplier:   VWR Collection
VWR®, 臨床用4ウェイチューブラック、
Supplier:  VWR Collection
Certificates
VWR®, 臨床用4ウェイチューブラック、
VWR®, 臨床用4ウェイチューブラック、
5 Options Available Below
Product Details & Documents
These tube racks have a six way connection to create various configurations and are designed to hold round, flat and conical blood collection and culture tubes.
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezer
  • Accommodates all brands of 13 and 17 mm tubes
They can be used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed.
Product Family Options
Product Information
  • 説明
    Colour
    穴の数
    For tubes
    Availability
    Price
  • Clinical 4-way tube racks
    Purple
    12/18/12/18
    13/17 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Clinical 4-way tube racks
    Assorted
    12/18/12/18
    13/17 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Clinical 4-way tube racks
    Blue
    12/18/12/18
    13/17 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Clinical 4-way tube racks
    Green
    12/18/12/18
    13/17 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
  • Clinical 4-way tube racks
    Natural
    12/18/12/18
    13/17 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items