VWR® Mr. Lab 2.0, 実験用椅子、
Supplier: VWR Collection
Certificates
About this item
Mr. Lab 2.0 features its own laboratory ergonomics. Its hygienic upholstery is made of artificial leather. Innovative airflow technology provides ideal climate comfort. Smart details such as the soft grip function handle, an integrated hook, and a labelling field are practical and improve safety in the laboratory.
- Ideal climate comfort thanks to the innovative airflow technology
- Perfect laboratory ergonomics
- Auto-synchron technology with automatic weight regulation
- Seat tilt and seat depth adjustment
- Soft grip handle (for more mobility, protects the upholstery from contamination)
- Integrated hook (e.g. for bag, backpack, lab coats)
- Clever labelling field (enables assignment to application areas and personalisation)
- Artificial leather: Easy to clean, resistant to disinfectants, soft and comfortable
- Hygienic design
- Extremely robust
- Optional 4D armrests
All-inclusive ergonomics with auto-synchron technology, automatic weight regulation, seat height adjustment, backrest height adjustment by 60 mm, seat tilt adjustment by 6°, seat depth adjustment by 60 mm, solid aluminium polished star base.
Certifications: GS certified, suitable for use in biosafety laboratories BSL 1 and BSL 2. GMP conformity.
Ordering information: 10 year warranty.
Specifications
- Catalog No:
- 132-1412
- 132-1411
- 132-1410
- Environmentally Preferable:
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Environmental certification:
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)