Orders LinkContactUsLinkComponent
VWR®, 万能フィルターチップ
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 万能フィルターチップ
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 万能フィルターチップ
9 Options Available Below
Product Details & Documents
VWR® Universal low retention filter tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with the widest range of pipette
    Maximises sample recovery with low binding resin and a fine point design
    Improves ergonomics by reducing the force required to attach and eject a tip
    Sterility Assurance Level (SAL) of 10-3
    Autoclavable at 1 bar, 122 °C for 15 minutes
    Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to 10 times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
 : Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
Specifications for Product Family
Specifications
  • Catalog No:
  • 732-3709
  • 732-3630
  • 732-3631
  • 732-3632
  • 732-3633
  • 732-3634
  • 732-3636
  • 732-3637
  • 732-3638
  • Environmentally Preferable:
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
Specifications
Product Family Options
Product Information
  • Tip style
    容量
    Sterile
    Filter
    長さ
    Colour code
    Packed
    Availability
    Price
  • Universal, low retention
    2 µl
    +
    +
    31,24 mm
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3709
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    10 µl
    +
    +
    31,24 mm
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3630
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, extended, low retention
    10 µl
    +
    +
    45,84 mm
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3631
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    20 µl
    +
    +
    50,75 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3632
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    100 µl
    +
    +
    59,54 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3633
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    200 µl
    +
    +
    59,54 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3634
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    300 µl
    +
    +
    59,54 mm
    Green
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3636
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    1000 µl
    +
    +
    81,82 mm
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3637
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • Universal, low retention
    1250 µl
    +
    +
    98,40 mm
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    732-3638
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging