Orders LinkContactUsLinkComponent
VWR®, LTS®対応ピペットチップ
VWR®, LTS®対応ピペットチップ
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, LTS®対応ピペットチップ
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, LTS®対応ピペットチップ
20 Options Available Below
Product Details & Documents
VWR® LTS® compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS® compatible pipettors.
  • Compatible with the LTS® style pipettors
  • Maximises sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 1 bar, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
Caution: Reloads will only fit into empty racks 613-6445 or 613-6446, respectively.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-6507
  • 613-6512
  • 613-6495
  • 613-6502
  • 613-6508
  • 613-6513
  • 613-6496
  • 613-6503
  • 613-6509
  • 613-6514
  • 613-6497
  • 613-6504
  • 613-6510
  • 613-6515
  • 613-6498
  • 613-6505
  • 613-6511
  • 613-6516
  • 613-6499
  • 613-6506
  • Environmentally Preferable:
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
Specifications
Product Family Options
Product Information
  • 容量
    Sterile
    Filter
    長さ
    Colour code
    使用対象
    Packed
    Availability
    Price
  • 20 µl
    -
    -
    49,63 mm
    Red
    Rainin LTS style pipettes
    10 racks, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6507
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 20 µl
    +
    -
    49,63 mm
    Red
    Rainin LTS style pipettes
    10 racks, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6512
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 20 µl
    -
    -
    49,63 mm
    Red
    Rainin LTS style pipettes
    10 reloads, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6495
    Environmentally Preferable
    Environmental certification
  • 20 µl
    +
    -
    49,63 mm
    Red
    Rainin LTS style pipettes
    10 reloads, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6502
    Environmentally Preferable
    Environmental certification
  • 250 µl
    -
    -
    50,75 mm
    Green
    Rainin LTS style pipettes
    10 racks, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6508
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 250 µl
    +
    -
    50,75 mm
    Green
    Rainin LTS style pipettes
    10 racks, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6513
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 250 µl
    -
    -
    50,75 mm
    Green
    Rainin LTS style pipettes
    10 reloads, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6496
    Environmentally Preferable
    Environmental certification
  • 250 µl
    +
    -
    50,75 mm
    Green
    Rainin LTS style pipettes
    10 reloads, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6503
    Environmentally Preferable
    Environmental certification
  • 300 µl
    -
    -
    58,93 mm
    Yellow
    Rainin LTS style pipettes
    10 racks, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6509
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 300 µl
    +
    -
    58,93 mm
    Yellow
    Rainin LTS style pipettes
    10 racks, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6514
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 300 µl
    -
    -
    58,93 mm
    Yellow
    Rainin LTS style pipettes
    10 reloads, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6497
    Environmentally Preferable
    Environmental certification
  • 300 µl
    +
    -
    58,93 mm
    Yellow
    Rainin LTS style pipettes
    10 reloads, 96/pack, 5 packs/case
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6504
    Environmentally Preferable
    Environmental certification
  • 1000 µl
    -
    -
    88,90 mm
    Blue
    Rainin LTS style pipettes
    10 racks, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6510
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1000 µl
    +
    -
    88,90 mm
    Blue
    Rainin LTS style pipettes
    8 racks, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6515
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1000 µl
    -
    -
    88,90 mm
    Blue
    Rainin LTS style pipettes
    10 reloads, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6498
    Environmentally Preferable
    Environmental certification
  • 1000 µl
    +
    -
    88,90 mm
    Blue
    Rainin LTS style pipettes
    10 reloads, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6505
    Environmentally Preferable
    Environmental certification
  • 1200 µl
    -
    -
    97,66 mm
    Grey
    Rainin LTS style pipettes
    10 racks, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6511
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1200 µl
    +
    -
    97,66 mm
    Grey
    Rainin LTS style pipettes
    10 racks, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6516
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1200 µl
    -
    -
    97,66 mm
    Grey
    Rainin LTS style pipettes
    10 reloads, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6499
    Environmentally Preferable
    Environmental certification
  • 1200 µl
    +
    -
    97,66 mm
    Grey
    Rainin LTS style pipettes
    10 reloads, 96/pack, 4 packs/case
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6506
    Environmentally Preferable
    Environmental certification
Empty racks
Product Information
  • Catalog #
    Colour
    説明
    Availability
    Price
  • Transparent
    Empty racks for 10 µl to 300 µl tip reloads, universal and LTS® compatible, autoclavable
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - $0.00
    $0.00
    1 * 10 items
       $0.00
Product Information
  • Catalog #
    Colour
    説明
    Availability
    Price
  • Transparent
    Empty racks for 1000 µl and 1250 µl tip reloads, universal and LTS® compatible, autoclavable
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - $0.00