Orders LinkContactUsLinkComponent
VWR®, 万能ピペットチップ
VWR®, 万能ピペットチップ
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 万能ピペットチップ
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 万能ピペットチップ
33 Options Available Below
Product Details & Documents
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with the widest range of pipette
  • Maximises sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
注意: Reloads will fit into emptied racks of respective racked tip version or into the empty racks 613-6445 or 613-6446, respectively.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-6462
  • 613-6453
  • 613-6440
  • 613-6433
  • 613-6416
  • 613-6463
  • 613-6454
  • 613-6441
  • 613-6434
  • 613-6417
  • 613-6465
  • 613-6456
  • 613-6442
  • 613-6435
  • 613-6418
  • 613-6468
  • 613-6458
  • 613-6436
  • 613-6419
  • 613-6469
  • 613-6459
  • 613-6443
  • 613-6437
  • 613-6420
  • 613-6470
  • 613-6460
  • 613-6438
  • 613-6421
  • 613-6471
  • 613-6461
  • 613-6444
  • 613-6439
  • 613-6422
  • Environmentally Preferable:
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Waste Reducing|Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
Specifications
Product Family Options
10 µl
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 10 µl
    +
    31,24 mm
    Natural
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6462
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 10 µl
    -
    31,24 mm
    Natural
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6453
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 10 µl
    +
    31,24 mm
    Natural
    Red
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6440
    Environmentally Preferable
    Environmental certification
  • 10 µl
    -
    31,24 mm
    Natural
    Red
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6433
    Environmentally Preferable
    Environmental certification
  • 10 µl
    -
    31,24 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6416
    Environmentally Preferable
    Environmental certification
10 µl extended
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 10 µl
    +
    45,84 mm
    Natural
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6463
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 10 µl
    -
    45,84 mm
    Natural
    Red
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6454
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 10 µl
    +
    45,84 mm
    Natural
    Red
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6441
    Environmentally Preferable
    Environmental certification
  • 10 µl
    -
    45,84 mm
    Natural
    Red
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6434
    Environmentally Preferable
    Environmental certification
  • 10 µl
    -
    45,84 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6417
    Environmentally Preferable
    Environmental certification
200 µl
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 200 µl
    +
    50,75 mm
    Natural
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6465
    Environmentally Preferable
    Environmental certification
  • 200 µl
    -
    50,75 mm
    Natural
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6456
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 200 µl
    +
    50,75 mm
    Natural
    Yellow
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6442
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 200 µl
    -
    50,75 mm
    Natural
    Yellow
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6435
    Environmentally Preferable
    Environmental certification
  • 200 µl
    -
    50,75 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6418
    Environmentally Preferable
    Environmental certification
250 µl
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 250 µl
    +
    59,54 mm
    Natural
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6468
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 250 µl
    -
    59,54 mm
    Natural
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6458
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 250 µl
    -
    59,54 mm
    Natural
    Yellow
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6436
    Environmentally Preferable
    Environmental certification
  • 250 µl
    -
    59,54 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6419
    Environmentally Preferable
    Environmental certification
300 µl
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 300 µl
    +
    59,54 mm
    Natural
    Green
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6469
    Environmentally Preferable
    Environmental certification
  • 300 µl
    -
    59,54 mm
    Natural
    Green
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6459
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 300 µl
    +
    59,54 mm
    Natural
    Green
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6443
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 300 µl
    -
    59,54 mm
    Natural
    Green
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6437
    Environmentally Preferable
    Environmental certification
  • 300 µl
    -
    59,54 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6420
    Environmentally Preferable
    Environmental certification
1000 µl
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 1000 µl
    +
    81,82 mm
    Natural
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6470
    Environmentally Preferable
    Environmental certification
  • 1000 µl
    -
    81,82 mm
    Natural
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6460
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1000 µl
    -
    81,82 mm
    Natural
    Blue
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6438
    Environmentally Preferable
    Environmental certification
  • 1000 µl
    -
    81,82 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6421
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
1250 µl
Product Information
  • 容量
    Sterile
    長さ
    Tip colour
    Colour code
    Packed
    Availability
    Price
  • 1250 µl
    +
    98,40 mm
    Natural
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6471
    Environmentally Preferable
    Environmental certification
  • 1250 µl
    -
    98,40 mm
    Natural
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6461
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1250 µl
    +
    98,40 mm
    Natural
    Blue
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
      Waste Reducing|Sustainable Packaging
    More
    Specifications
    Cat. No.
    613-6444
    Environmentally Preferable
    Waste Reducing|Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing|Sustainable Packaging
  • 1250 µl
    -
    98,40 mm
    Natural
    Blue
    10 reloads, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6439
    Environmentally Preferable
    Environmental certification
  • 1250 µl
    -
    98,40 mm
    Natural
    Bulk, 1000 tips/bag
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    613-6422
    Environmentally Preferable
    Environmental certification
Empty racks
Product Information
  • Catalog #
    Colour
    説明
    Availability
    Price
  • Transparent
    Empty racks for 1000 µl and 1250 µl tip reloads, universal and LTS® compatible, autoclavable
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - $0.00
    $0.00
    1 * 10 items
       $0.00
Product Information
  • Catalog #
    Colour
    説明
    Availability
    Price
  • Transparent
    Empty racks for 10 µl to 300 µl tip reloads, universal and LTS® compatible, autoclavable
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - $0.00