Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
CyMet
CyMet 22 for hematology CN-free, J.T.Baker®
CyMet 22 for hematology CN-free, J.T.Baker®
2986.0500PE
:
Avantor
:
CyMet 22 for hematology CN-free, J.T.Baker®
2986.0500PE
:
Avantor
:
Pk:
500 mL
Seq:
2725
Conductivity (mS):
24.9 - 25.9
Osmolality:
504 - 516 mOsm/kg
Performance Test:
passes test
pH at 25°C:
4.77 - 5.07
:
:
Refrigerator
:
1