Orders LinkContactUsLinkComponent
ガラスマイクロファイバーフィルタ、RankemTM
ガラスマイクロファイバーフィルタ、RankemTM
Supplier:   Transene
ガラスマイクロファイバーフィルタ、RankemTM
Supplier:  Transene
Certificates
ガラスマイクロファイバーフィルタ、RankemTM
3 Options Available Below
Product Details & Documents
Glass microfiber filters are made of 100% borosilicate glass, and it is highly pure and extremely white in colour. Fast flow rates with high load capacity and high retention of fine particles. It can even withstand temperature up to 500 °C.
These Glass microfiber filters find application in:
  • Air pollution monitoring and estimation of the suspended particles in air
  • As a pre filter in highly coarse sample in HPLC analysis ideal for pre filtration to enhance sample volume of syringe filters
  • Removal of precipitates in ion-pair reagents
  • Clarification of reagents and solutions in spectrophotometry
  • Determination of sediment in petroleum products
These are completely binder free, hence no leaching and no contamination of filtrate, biologically inert, non hygroscopic.
Product Family Options
Product Information
  • Grade
    サイズ
    Availability
    Price
  • GMF sheet, Grade A
    460×570 mm
    1 * 1 items
  • GMF sheet, Grade A
    203×254 mm
    1 * 1 items
  • GMF thimble, Grade A
    19×90 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items