Rankem®, Serological Pipettes, Graduated, Class A
: Transene
Borosilicate glass pipettes, class A.
- Colour coded
- Graduated upto tip
- Calibrated to deliver their total capacity
: Supplied with certificate.
Frequently Bought Together
Borosilicate glass pipettes, class A.