Orders LinkContactUsLinkComponent
細菌培養用ピペット、Rankem™(ランケム)
細菌培養用ピペット、Rankem™(ランケム)
Supplier:   Transene
細菌培養用ピペット、Rankem™(ランケム)
Supplier:  Transene
Certificates
細菌培養用ピペット、Rankem™(ランケム)
2 Options Available Below
Product Details & Documents
Made of grade 3.3 borosilicate glass.
  • Graduated
Product Family Options
Product Information
  • 容量
    Tolerance
    Availability
    Price
  • 1,1 ml
    ± 0,025
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • 2,2 ml
    ± 0,040