Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
ボトル
Glass Bottles
Rankem®, Reagent Bottles, with Screw Cap
Rankem®, Reagent Bottles, with Screw Cap
Rankem®, Reagent Bottles, with Screw Cap
Catalog # RANKBR-WMSC-00030
Supplier:
Transene
Expand image
Rankem®, Reagent Bottles, with Screw Cap
Catalog # RANKBR-WMSC-00030
Supplier:
Transene
Specifications
容量:
30 ml
Seq:
01
Pk:
20
Specifications
About this item
Product Details
Made of grade 3.3 borosilicate glass.
Graduated
Product Details