Orders LinkContactUsLinkComponent
Zorbaxシリカ, HPLCカラム
Zorbaxシリカ, HPLCカラム
   Sustainable Options Available
Supplier:   HICHROM
Zorbaxシリカ, HPLCカラム
Supplier:  HICHROM
   Sustainable Options Available
Certificates
Zorbaxシリカ, HPLCカラム
12 Options Available Below
Product Details & Documents
Zorbax spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerisation process.
  • Spherical porous silica
  • Narrow particles and pore size distribution
  • Robust with good batch to batch reproducibility
The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
Ordering information: Other dimensions columns and guard cartridges are available on request.
Specifications for Product Family
Specifications
  • Catalog No:
  • ZSIL-50AM
  • ZSIL-100AM
  • ZSIL-150AM
  • ZSIL-250AM
  • ZSIL-50AS
  • ZSIL-100AS
  • ZSIL-150AS
  • ZSIL-250AS
  • ZSIL-50A
  • ZSIL-100A
  • ZSIL-10CM5
  • ZSIL-10C5-SK
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Surface area:
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Family Options
Zorbax microbore columns
Product Information
  • Phase
    カラム形式
    細孔サイズ
    粒子サイズ
    内径
    長さ
    Availability
    Price
  • Silica
    70 Å
    5 µm
    2,10 mm
    50 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-50AM
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    2,10 mm
    100 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-100AM
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    2,10 mm
    150 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-150AM
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    2,10 mm
    250 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-250AM
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
Zorbax medium bore columns
Product Information
  • Phase
    カラム形式
    細孔サイズ
    粒子サイズ
    内径
    長さ
    Availability
    Price
  • Silica
    70 Å
    5 µm
    3,20 mm
    50 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-50AS
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    3,20 mm
    100 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-100AS
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    3,20 mm
    150 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-150AS
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    3,20 mm
    250 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-250AS
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
Zorbax analytical columns
Product Information
  • Phase
    カラム形式
    細孔サイズ
    粒子サイズ
    内径
    長さ
    Availability
    Price
  • Silica
    70 Å
    5 µm
    4,60 mm
    50 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-50A
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Silica
    70 Å
    5 µm
    4,60 mm
    100 mm
    1 * 1 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-100A
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
Zorbax guard cartridges
Product Information
  • Phase
    カラム形式
    細孔サイズ
    粒子サイズ
    内径
    長さ
    Availability
    Price
  • Silica
    Analytical
    70 Å
    5 µm
    2,10 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-10CM5
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
Zorbax guard cartridge starter kit
Product Information
  • Phase
    カラム形式
    細孔サイズ
    粒子サイズ
    内径
    長さ
    Availability
    Price
  • Silica
    Analytical
    70 Å
    5 µm
    3,20 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    ZSIL-10C5-SK
    Environmentally Preferable
    Low Manufacturing Impact
    Endcapped
    No
    Surface area
    300 m²/g
    Environmental certification
    ISO 14001:2015 (530840 UM15)