Rankem®, Volumetric Flasks, Class B, Glass, clear, Interchangeable Stopper
: Transene
Volumetric flask with interchangeable stopper with accuracy as per Class B.
- Borosilicate glass
- Volumetric flasks 1 and 2 ml are of test tube shape
Frequently Bought Together