Orders LinkContactUsLinkComponent
真空デシケーター、Rankem™ (ランケム)
真空デシケーター、Rankem™ (ランケム)
Supplier:   Transene
真空デシケーター、Rankem™ (ランケム)
Supplier:  Transene
Certificates
真空デシケーター、Rankem™ (ランケム)
4 Options Available Below
Product Details & Documents
These Rankem™ desiccators are made of soft glass.
  • With tubulated cover
  • Feature stopper with PTFE spindle
Product Family Options
Product Information
  • 説明
    内径
    Availability
    Price
  • Desiccator
    150 mm
    1 * 1 items
  • Desiccator
    200 mm
    1 * 1 items
  • Desiccator
    250 mm
    1 * 1 items
  • Desiccator
    300 mm