Orders LinkContactUsLinkComponent
J.T.Baker®, Hematology Controls, 3-diff Control Extended
Supplier:   Avantor
J.T.Baker®, Hematology Controls, 3-diff Control Extended
Supplier:  Avantor
Certificates
J.T.Baker®, Hematology Controls, 3-diff Control Extended
3 Options Available Below
Product Details & Documents
In vitro diagnostic hematology control product, intended for professional use only.
 : Parameter Controls are to be stored upright at 2-8°C when not in use. Stored at this temperature, the Controls are guaranteed stable until the expiry date.
Product Family Options
Product Information
  • 説明
    Pk
    Availability
    Price
  • 3-Different control extended, low
    2,5 mL
    Log in for availability
    Log in for availability
    1 * 2,5 mL
    1 * 2,5 mL
  • 3-Different control extended, medium
    2,5 mL
    Log in for availability
    Log in for availability
    1 * 2,5 mL
    1 * 2,5 mL
  • 3-Different control extended, high
    2,5 mL
    Log in for availability
    Log in for availability
    1 * 2,5 mL
    1 * 2,5 mL