Orders LinkContactUsLinkComponent
ピペット用バルブ, ランケム
Supplier:   Transene
ピペット用バルブ, ランケム
Supplier:  Transene
Certificates
ピペット用バルブ, ランケム
3 Options Available Below
Product Details & Documents
These pipette bulbs are made of silicone.
 : Packet contains 2 small, 2 medium and 1 large pipette bulbs.
Product Family Options
Product Information
  • 説明
    容量
    Colour
    Availability
    Price
  • Pipette bulb, small
    5 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Pipette bulb, medium
    10 ml
    Red
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Pipette bulb, large
    15 ml
    Red