Orders LinkContactUsLinkComponent
シリンジフィルター、RanDisc™、ナイロン
シリンジフィルター、RanDisc™、ナイロン
Supplier:   Transene
シリンジフィルター、RanDisc™、ナイロン
Supplier:  Transene
Certificates
シリンジフィルター、RanDisc™、ナイロン
2 Options Available Below
Product Details & Documents
RanDisc syringe filter with nylon membrane are ready to use syringe filters which provides high flow rates at low inlet pressures. High flow rates also ensures high total throughputs i.e. filtration requirements with volumes upto 10 to 15 ml to be clarified or ultracleaned.
  • Simple, rapid and reliable ultra cleaning of small volume samples for HPLC/GC analysis
  • For rapid small volume sterilisation
  • For particle removal, ultra cleaning and purification
Product Family Options
Product Information
  • 直径
    細孔サイズ
    Membrane
    Availability
    Price
  • 13 mm
    0,22 µm
    Nylon
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • 25 mm
    0,45 µm