Rankem® RanDisc™, Syringe Filters, PTFE Membrane
Catalog # RANKPT0225SF
Supplier: Transene
Specifications
- 直径:25 mm
- Membrane:PTFE
- 細孔サイズ:0,22 µm
- Seq:03
- Pk:50
Specifications
About this item
These syringe filters are ideal for rapid and reliable ultra-cleaning of small volume samples for HPLC or GC analysis, which require an even more chemical resistant unit for solvents such as acetone dimethylformamide and DMSO or for aggressive aqueous liquids.
- Ready to use units
- Hydrophobic