Orders LinkContactUsLinkComponent
Rankem®, Transfer Pipettes, Class A
Rankem®, Transfer Pipettes, Class A
Supplier:   Transene
Rankem®, Transfer Pipettes, Class A
Supplier:  Transene
Certificates
Rankem®, Transfer Pipettes, Class A
10 Options Available Below
Product Details & Documents
Transfer pipettes has accuracy as per Class A (with certificate).
  • Volumetric
  • One mark
  • Borosilicate glass
Product Family Options
Product Information
  • 説明
    容量
    Availability
    Price
  • Transfer pipettes, tolerance: ±0,007 ml
    1 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,01 ml
    2 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,01 ml
    3 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,01 ml
    4 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,015 ml
    5 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,02 ml
    10 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,03 ml
    20 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,03 ml
    25 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,05 ml
    50 ml
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Transfer pipettes, tolerance: ±0,08 ml
    100 ml