Orders LinkContactUsLinkComponent
VWR® Traceable® Precision Sentry™, Thermometer with Probe
VWR® Traceable® Precision Sentry™, Thermometer with Probe
Supplier:   VWR Collection
VWR® Traceable® Precision Sentry™, Thermometer with Probe
Supplier:  VWR Collection
Certificates
VWR® Traceable® Precision Sentry™, Thermometer with Probe
VWR® Traceable® Precision Sentry™, Thermometer with Probe
3 Options Available Below
Product Details & Documents
Simple one-key reset operation resets MIN/MAX memories.
  • Triple display constantly shows high, low, and current temperatures
  • Sensor and 10-foot cable may be placed underwater, in refrigerator, or freezer
  • Unplugging the cable activates a sensor inside the unit for monitoring ambient temperature
Simultaneously displays high/low and current temperature. Instantly view if freezer, water bath, or incubator is within temperature spec. Monitor temperature of any experiment when you’re not there—overnight, weekends, even during lunch. Small cable diameter (1/16") allows refrigerator doors to close on it.
Simple, one key operation resets the minimum/maximum memories. Probe’s wire-mounting bracket permits easy sensor placement. Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Unit’s high-impact ABS plastic case has a flip-open stand, wall mount, magnet, and hook and loop tape mounting tape.
Certifications: CE marked.
Ordering information: Supplied with Traceable® Certificate, flip-open stand, wall mount, magnet, and hook and loop tape.
Specifications for Product Family
Specifications
  • Catalog No:
  • 620-2718
  • 620-2719
  • 620-2720
  • W×D×H:
  • 2¾×2¼×½"
  • 2¾×2¼×½"
  • 2¾×2¼×½"
Specifications
Product Family Options
Product Information
  • Range
    Accuracy
    解像度
    Probe
    Weight
    Availability
    Price
  • –50,0...+60,0 °C (–58,0...140,0 °F)
    ±0,5 °C (−20...+60 °C), 1 °C otherwise
    0,1 °C
    Bullet
    17/8 oz.
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    620-2718
    W×D×H
    2¾×2¼×½"
  • –50,0...+60,0 °C (–58,0...140,0 °F)
    ±0,5 °C (−20...+60 °C), 1 °C otherwise
    0.1°
    Bottle
    17/8 oz.
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    620-2719
    W×D×H
    2¾×2¼×½"
  • –50,0...+60,0 °C (–58,0...140,0 °F)
    ±0,5 °C (−20...+60 °C), 1 °C otherwise
    0.1°
    Vaccine bottle
    17/8 oz.
    1 * 1 items
    Specifications:
    More
    Specifications
    Cat. No.
    620-2720
    W×D×H
    2¾×2¼×½"